Learn More
Electron Microscopy Sciences Corning Digital Stirring Hot Plate Two 9" Support Rods
SDP
Catalog No. 501899845
Encompass_Preferred
Two 9" support rods that screw together, use with models: PC-400/410/420, PC-400D-410D/ 420D, PC-600D/610D/620D.
Use: 9"
Catalog No. 50-189-9845 Supplier Electron Microscopy Sciences Supplier No. 7235125
Two 9" support rods that screw together, use with models: PC-400/410/420, PC-400D-410D/ 420D, PC-600D/610D/620D.
Use: 9"