Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Support Rod
SDP

Catalog No. 11392168
Change view
Click to view available options
Length (Metric):
470 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-392-168 470 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-168 Supplier Velp™ Supplier No. A00001069
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use With: Velp Heating Magnetic Stirrer and Hot Plate

For Use With: Velp Heating Magnetic Stirrer and Hot Plate

  • For use with AREX™ CerAlTop™ Hot Plate Stirrers
  • Helps keep round-bottom flasks uprigth during stirring/heating

Specifications

Diameter (Metric) 12 mm
For Use With (Equipment) Velp Heating Magnetic Stirrer and Hot Plate
Length (Metric) 470 mm
Type Support Rod
Warranty 24 Months
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.