Learn More
Bellco Glass, Inc. Support Rubber Cushion,600mL For 3045-00600 Tube
SDP
Catalog No. 501945589
Encompass_Preferred
Support cushion is used for insertion into centrifuge bucket under glass centrifuge tube to prevent breakage, adds stability For 3045-00600 tube.
Catalog No. 50-194-5589 Supplier Bellco Glass, Inc. Supplier No. 304510600
Support cushion is used for insertion into centrifuge bucket under glass centrifuge tube to prevent breakage, adds stability For 3045-00600 tube.