Learn More
Hach Company Support, Separatory Funnel
Supplier:  Hach Company 2238600
Encompass
Separatory Funnel Support made of Polypropylene and molded in one piece. Chuck fits rods with diameters from 8 mm to 14 mm. Ring is tapered 72-85 mm diameter.
Catalog No. 50-199-5595