Learn More
Ace Glass, Inc. Lab stand support shelf, 7x10 inches, no hole, fits lab support rod up to 5/8 inch
SDP
Catalog No. 50977220
Encompass
7 x 10 lab stand support shelf fits any stand with up to 5/8 O.D. rod. Manufactured from light, cast aluminum and epoxy coated.
Catalog No. 50-977-220 Supplier Ace Glass, Inc. Supplier No. 1117308
7 x 10 lab stand support shelf fits any stand with up to 5/8 O.D. rod. Manufactured from light, cast aluminum and epoxy coated.