Learn More
Chemglass Life Sciences Support, Solid Tripod, 100mm Leg Length
SDP
Catalog No. 501311214
Encompass_Preferred
Support, Solid Tripod, 100mm Leg Length. Base is made of cast iron with a baked black finish. Rod is 10mm O.D. x 600mm (24”). Available in two sizes.
Catalog No. 50-131-1214 Supplier Chemglass Life Sciences Supplier No. CG3058200
Support, Solid Tripod, 100mm Leg Length. Base is made of cast iron with a baked black finish. Rod is 10mm O.D. x 600mm (24”). Available in two sizes.