Learn More
Chemglass Life Sciences Support, Solid Tripod, 150mm Leg Length
SDP
Catalog No. 501311215
Encompass_Preferred
Support, Solid Tripod, 150mm Leg Length. Base is made of cast iron with a baked black finish. Rod is 10mm O.D. x 600mm (24”). Available in two sizes.
Catalog No. 50-131-1215 Supplier Chemglass Life Sciences Supplier No. CG3058205
Support, Solid Tripod, 150mm Leg Length. Base is made of cast iron with a baked black finish. Rod is 10mm O.D. x 600mm (24”). Available in two sizes.