Learn More
BURKLE INC Support stand for two compact jerrycans 1.32 gal. (5l)
Catalog No. 5000025646
Encompass_Preferred
The support is designed to securely and space-efficiently fix 5-liter canisters, preventing slipping, tipping, and concentrated load points. Made of lightweight, rustproof aluminum.
Two tall tabs enclose the narrow canister sides, preventing shifting or tipping, ideal for shelves, vehicles, or vibrating environments like labs and workshops.
The base plate evenly distributes the full canister’s weight, protecting surfaces, reducing pressure points, preventing deformation, and easing removal.
By locking multiple canisters side by side, the system enables space-optimized storage, maximizing cabinet or shelf capacity like tightly arranged books.
Catalog No. 50-000-25646 Supplier BURKLE INC Supplier No. 04359102
The support is designed to securely and space-efficiently fix 5-liter canisters, preventing slipping, tipping, and concentrated load points. Made of lightweight, rustproof aluminum.
Two tall tabs enclose the narrow canister sides, preventing shifting or tipping, ideal for shelves, vehicles, or vibrating environments like labs and workshops.
The base plate evenly distributes the full canister’s weight, protecting surfaces, reducing pressure points, preventing deformation, and easing removal.
By locking multiple canisters side by side, the system enables space-optimized storage, maximizing cabinet or shelf capacity like tightly arranged books.