Learn More
World Precision Instrument Support Stand for Ussing System
SDP
Catalog No. 502521264
Encompass
Ussing system stand
This stand includes two reservoir mounting rods and clamps, and a chamber mount with adjustable screws. The Ussing system has been discontinued, and these replacement parts are available while supplies last.
Catalog No. 50-252-1264 Supplier World Precision Instrument Supplier No. 5153
Ussing system stand
This stand includes two reservoir mounting rods and clamps, and a chamber mount with adjustable screws. The Ussing system has been discontinued, and these replacement parts are available while supplies last.