Learn More
Sigma Aldrich Fine Chemicals Biosciences Support stand with rod bas1EA
Catalog No. 502901017
Encompass
Base is steel with black enamel finish. Removable rod is zinc-plated steel. Heavy-duty styles have an added steel plate on the back for extra weight.
Catalog No. 50-290-1017 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5094421EA
Base is steel with black enamel finish. Removable rod is zinc-plated steel. Heavy-duty styles have an added steel plate on the back for extra weight.