Learn More
Chemglass Life Sciences Support Tubes, Stainless Steel, 1/2in OD X 60in Length
SDP
Catalog No. 50873650
Encompass_Preferred
Support Tubes, Stainless Steel, 1/2in OD X 60in Length. These support tubes are made of brilliant polished 304 stainless steel. They have good surface hardness and do not score from clamp binding screws. Tubes are 1/2" (13mm) in diameter, and are hollow, reducing weight to 1/2 of solid stainless steel rods while maintaining all the advantages of the stainless steel. The walls are 1/16" (1.6mm) thick, strong, and do not sag. The ends of each tube are capped with polyethylene tight fitting end-caps.
Catalog No. 50-873-650 Supplier Chemglass Life Sciences Supplier No. CG305107
Support Tubes, Stainless Steel, 1/2in OD X 60in Length. These support tubes are made of brilliant polished 304 stainless steel. They have good surface hardness and do not score from clamp binding screws. Tubes are 1/2" (13mm) in diameter, and are hollow, reducing weight to 1/2 of solid stainless steel rods while maintaining all the advantages of the stainless steel. The walls are 1/16" (1.6mm) thick, strong, and do not sag. The ends of each tube are capped with polyethylene tight fitting end-caps.