Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 1 Chromium-Free Steel
SDP
Catalog No. 501534373
Encompass
Lateral support walls made of chromium-free steel for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: Chromium-free steel
Abrasion resistance: fairly good
Suitable for: medium hard, brittle samples
Catalog No. 50-153-4373 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0071.09
Lateral support walls made of chromium-free steel for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: Chromium-free steel
Abrasion resistance: fairly good
Suitable for: medium hard, brittle samples