Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 1 Stainless Steel
SDP
Catalog No. 501534374
Encompass
Lateral support walls made of stainless steel for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples
Catalog No. 50-153-4374 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0080.10
Lateral support walls made of stainless steel for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples