Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 1 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534375
Encompass
Lateral support walls made of hardmetal tungsten carbide for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: extremely good
Suitable for: hard, abrasive samples
Catalog No. 50-153-4375 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0090.08
Add to Cart
Add to Cart
Request a Quote
Lateral support walls made of hardmetal tungsten carbide for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: extremely good
Suitable for: hard, abrasive samples