Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 1 Hardened Steel
SDP
Catalog No. 501534372
Encompass
Lateral support walls made of hardened steel belong to the standard equipment of the FRITSCH Jaw Crushers for PULVERISETTE 1 classic line, Size 1 and are therefore included in the instrument price.
General material specification:
Material: hardened steel - 1.2080
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4372 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0070.09
Lateral support walls made of hardened steel belong to the standard equipment of the FRITSCH Jaw Crushers for PULVERISETTE 1 classic line, Size 1 and are therefore included in the instrument price.
General material specification:
Material: hardened steel - 1.2080
Abrasion resistance: good
Suitable for: hard, brittle samples