Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 2 Chromium-Free Steel
SDP
Catalog No. 501534391
Encompass
Lateral support walls made of chronium-free steel for FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2.
General material specifications:
Material:chromium-free steel
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4391 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3071.09
Lateral support walls made of chronium-free steel for FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2.
General material specifications:
Material:chromium-free steel
Abrasion resistance: good
Suitable for: hard, brittle samples