Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 2 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534393
Encompass
Lateral support walls made of hardmetal tungsten carbide for FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2.
General material specification:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: very good
Suitable for: hard, abrasive samples
Catalog No. 50-153-4393 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3090.08
Add to Cart
Add to Cart
Request a Quote
Lateral support walls made of hardmetal tungsten carbide for FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2.
General material specification:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: very good
Suitable for: hard, abrasive samples