Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 2 Zirconium Oxide
SDP
Catalog No. 501534399
Encompass
Lateral support walls made of zirconium oxide for FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2.
General material specifications:
Material: zirconium oxide
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4399 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3160.27
Add to Cart
Add to Cart
Request a Quote
Lateral support walls made of zirconium oxide for FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2.
General material specifications:
Material: zirconium oxide
Abrasion resistance: good
Suitable for: hard, brittle samples