Learn More
Fritsch Milling And Sizing Inc Support Walls P-1 Cl Size 2 Hardened Steel
SDP
Catalog No. 501534390
Encompass
Lateral support walls made of hardened steel belong to the standard equipment of the FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2 and therefore they are included in the instrument price.
General material specifications:
Material: hardened steel - 1.2080
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4390 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3070.09
Add to Cart
Add to Cart
Request a Quote
Lateral support walls made of hardened steel belong to the standard equipment of the FRITSCH Jaw Crusher PULVERISETTE 1 classic line Size 2 and therefore they are included in the instrument price.
General material specifications:
Material: hardened steel - 1.2080
Abrasion resistance: good
Suitable for: hard, brittle samples