Learn More
Thermco Products Inc SUPR PRE RTD PLAT THERM W/MEM
Catalog No. NC0995100
Encompass_Preferred
“SUPRA” PRECISION - RTD Platinum Thermometer w/ MEMORY & probe. Thermco’s CT375DIG is a very accurate temperature measuring systems. With the accuracy of 0.1 °C and resolution of 0.01°C, it will fulfill the most critical measuring requirement. The unit is equipped with stand and large backlight LCD that makes this unit perfect to setup as a test station on site.
Catalog No. NC0995100 Supplier Thermco Products Inc Supplier No. CT375DIG
Add to Cart
Add to Cart
“SUPRA” PRECISION - RTD Platinum Thermometer w/ MEMORY & probe. Thermco’s CT375DIG is a very accurate temperature measuring systems. With the accuracy of 0.1 °C and resolution of 0.01°C, it will fulfill the most critical measuring requirement. The unit is equipped with stand and large backlight LCD that makes this unit perfect to setup as a test station on site.