Learn More
Cytiva (Formerly Pall Lab) Supracap™ 50 depth filter capsule with dual-layer media grade PDP8 (6-30 m retention rating) and luer-lock connections
Supplier:  Cytiva SC050PDP8
Encompass
Supracap™ 50 depth filter capsule with dual-layer media grade PDP8 (6-30 m retention rating) and luer-lock connections
Catalog No. NC1557329