Learn More
Perkin Elmer US LLC Supra-Clean C18 SPE Column - 100 mg, 1 mL, Pkg. 100
Catalog No. 502104797
Encompass_Preferred
Recommended for extracting polar to non-polar compounds when analyzing toxins in food, drugs/drug metabolites in biological matrices, and trace organic materials in water.
Catalog No. 50-210-4797 Supplier Perkin Elmer US LLC Supplier No. N9306478
Recommended for extracting polar to non-polar compounds when analyzing toxins in food, drugs/drug metabolites in biological matrices, and trace organic materials in water.