Learn More
Perkin Elmer US LLC Supra-Clean C18 SPE Column Without End Cap - 500 mg, 3 mL, Pkg. 50
Catalog No. 502105084
Encompass_Preferred
Recommended for extracting polar to non-polar compounds when analyzing toxins in food, drugs/drug metabolites in biological matrices, and trace organic materials in water.
Catalog No. 50-210-5084 Supplier Perkin Elmer US LLC Supplier No. N9306642
Recommended for extracting polar to non-polar compounds when analyzing toxins in food, drugs/drug metabolites in biological matrices, and trace organic materials in water.