Learn More
Perkin Elmer US LLC Supra-Clean Silica SPE Column - 1 g, 6 mL, Pkg. 30
Catalog No. 502104749
Encompass_Preferred
Recommended for extracting polar compounds from aldehydes, amines, pesticides, herbicides, carotenoids, fat soluble vitamins, baflatoxins, fatty acids, and phospholipids.
Catalog No. 50-210-4749 Supplier Perkin Elmer US LLC Supplier No. N9306404
Recommended for extracting polar compounds from aldehydes, amines, pesticides, herbicides, carotenoids, fat soluble vitamins, baflatoxins, fatty acids, and phospholipids.