Learn More
Perkin Elmer US LLC Supra-Clean Silica SPE Column - 2 g, 15 mL, Pkg. 20
Catalog No. 502104850
Encompass_Preferred
Recommended for extracting polar compounds from aldehydes, amines, pesticides, herbicides, carotenoids, fat soluble vitamins, baflatoxins, fatty acids, and phospholipids.
Catalog No. 50-210-4850 Supplier Perkin Elmer US LLC Supplier No. N9306534
Recommended for extracting polar compounds from aldehydes, amines, pesticides, herbicides, carotenoids, fat soluble vitamins, baflatoxins, fatty acids, and phospholipids.