Learn More
Perkin Elmer US LLC Supra-Clean Strong Cation Exchange SPE Column - 200 mg, 3 mL, Pkg. 50
Catalog No. 502104880
Encompass_Preferred
Recommended for extracting basic compounds from cations, antibiotics, drugs, amino acids, catecholamines, herbicides, nucleic acid bases, nucleosides, and surfactants.
Catalog No. 50-210-4880 Supplier Perkin Elmer US LLC Supplier No. N9306538
Recommended for extracting basic compounds from cations, antibiotics, drugs, amino acids, catecholamines, herbicides, nucleic acid bases, nucleosides, and surfactants.