Learn More
Med Vet International Supracondylar Osteotomy Plate, 2.7mm Left, J1076A
Catalog No. 5000583766
Encompass_Preferred
Osteotomy of the distal femur is now a recognized technique for the management of patella luxation in medium and larger dogs with added femural abnormality.The use of straight plates is difficult due to hole positioning and plate contouring. These supra condylar plates are computer designed to fit this area. Pre-contoured to minimize intra operative adjustments.
Catalog No. 50-005-83766 Supplier Med Vet International Supplier No. JOR1076A
Osteotomy of the distal femur is now a recognized technique for the management of patella luxation in medium and larger dogs with added femural abnormality.The use of straight plates is difficult due to hole positioning and plate contouring. These supra condylar plates are computer designed to fit this area. Pre-contoured to minimize intra operative adjustments.