Learn More
Cayman Chemical Suprafenacine, Purity: ≥98%, CAS: 1477482-50-0, 5 mg
SDP
Catalog No. 502989449
Encompass_Preferred
An inhibitor of tubulin polymerization (IC50 = 0.38 μM) with anticancer activit; inhibits the proliferation of HeLa, Jurkat, SNU-16, MDA-MB-231, A549, SH-SY5Y, HCT15, and KB-3-1 cells (IC50s = 83-381.2 nM) and P-glycoprotein-enriched KB-VI cells (IC50 = 215.7 nM); increases phosphorylation of JNK and Bcl-2 and induces apoptosis and cell cycle arrest at the G2/M phase in HeLa cells at 10 μM; reduces tumor growth in an HCT15 colon adenocarcinoma mouse xenograft model at 20 mg/kg
Catalog No. 50-298-9449 Supplier Cayman Chemical Supplier No. 311585 MG
An inhibitor of tubulin polymerization (IC50 = 0.38 μM) with anticancer activit; inhibits the proliferation of HeLa, Jurkat, SNU-16, MDA-MB-231, A549, SH-SY5Y, HCT15, and KB-3-1 cells (IC50s = 83-381.2 nM) and P-glycoprotein-enriched KB-VI cells (IC50 = 215.7 nM); increases phosphorylation of JNK and Bcl-2 and induces apoptosis and cell cycle arrest at the G2/M phase in HeLa cells at 10 μM; reduces tumor growth in an HCT15 colon adenocarcinoma mouse xenograft model at 20 mg/kg