Learn More
Wilmad Labglass 4 mm Medium Wall Low Pressure/Vac Suprasil EPR Sample Tube 250 mm L
Catalog No. NC1571327
Encompass_Preferred
A 4X larger sealing surface eliminates leaks and greatly increases its lifetime when compared to traditional gas-tight tubes. It continues to offer a convenient flame-free sealing solution for air sensitive or volatile liquid samples at any pressure from vacuum to pressure slightly above 1 bar.
Catalog No. NC1571327 Supplier Wilmad Labglass Supplier No. 727LPV250M
A 4X larger sealing surface eliminates leaks and greatly increases its lifetime when compared to traditional gas-tight tubes. It continues to offer a convenient flame-free sealing solution for air sensitive or volatile liquid samples at any pressure from vacuum to pressure slightly above 1 bar.