Learn More
Electron Microscopy Sciences Superfrost Plus Gold Slide
SDP
Catalog No. 501899723
Encompass
These slides are specially designed for the application of fresh or formalin fixed frozen tissue. They are inherently tissue binding so there is no need to add sprays or adhesives to the slides.
Securely attach frozen breast, brain, skin and other normally difficult to bond tissue sections to the slide.
Ideal when special stains, immunocytochemical and in situ DNA hybridization techniques are applied on fresh and fixed frozen tissue.
Assist tissue adherence during immunocytochemical analysis of estrogen and progesterone receptors in fresh frozen breast tissue.
They are compatible with both toluidine blue and hematoxylin and eosin rapid frozen section stains.
Catalog No. 50-189-9723 Supplier Electron Microscopy Sciences Supplier No. 7186410
These slides are specially designed for the application of fresh or formalin fixed frozen tissue. They are inherently tissue binding so there is no need to add sprays or adhesives to the slides.
Securely attach frozen breast, brain, skin and other normally difficult to bond tissue sections to the slide.
Ideal when special stains, immunocytochemical and in situ DNA hybridization techniques are applied on fresh and fixed frozen tissue.
Assist tissue adherence during immunocytochemical analysis of estrogen and progesterone receptors in fresh frozen breast tissue.
They are compatible with both toluidine blue and hematoxylin and eosin rapid frozen section stains.