Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ SUPRO MAX™ Hand Cleaner Refill
SDP

Catalog No. 19043852 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
5000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-852 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-852 Supplier GOJO™ Supplier No. 757202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fast and effective heavy-duty hand cleaner with gentle scrubbers. Refill is gentle on skin and leaves them feeling healthy.

  • Fast and effective heavy-duty hand cleaner with gentle scrubbers
  • Unique blend of cleaners and skin conditioners help leave skin feeling healthy
  • Non-drying formula
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with each refill
  • Effective on oil, grease, paint, and adhesives
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Cleanser
Color Beige
Form Lotion
For Use With (Application) Effective on oil, grease, paint, and adhesives.
For Use With (Equipment) 7500-01
Quantity 2/Pk.
Size 5000 mL
Volume (Metric) 5000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.