Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Suprofen | 40828-46-4 | 99.1% | 50 MG
SDP

Catalog No. 502082439
Encompass_Preferred

Suprofen (TN-762) is a non-steroidal anti-inflammatory drug (NSAID) and an ibuprofen-type anti-inflammatory analgesic and antipyretic. It inhibits prostaglandin synthesis and has been proposed as an anti-arthritic. It is intended for research use only.

  • Non-steroidal anti-inflammatory drug (NSAID)
  • Inhibits prostaglandin synthesis
  • Purity of 99.1%
  • Molecular weight: 260.31
  • Chemical formula: C14H12O3S
  • Solid, white to off-white appearance
  • Recommended storage as powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO at ≥ 100 mg/mL
  • For research use only

Catalog No. 50-208-2439 Supplier Medchemexpress LLC Supplier No. HYB027050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suprofen (TN-762) is a non-steroidal anti-inflammatory drug (NSAID) and an ibuprofen-type anti-inflammatory analgesic and antipyretic. It inhibits prostaglandin synthesis and has been proposed as an anti-arthritic. It is intended for research use only.

  • Non-steroidal anti-inflammatory drug (NSAID)
  • Inhibits prostaglandin synthesis
  • Purity of 99.1%
  • Molecular weight: 260.31
  • Chemical formula: C14H12O3S
  • Solid, white to off-white appearance
  • Recommended storage as powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 2 years, -20°C for 1 year
  • Soluble in DMSO at ≥ 100 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.