Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Suprofen (TN-762) | 40828-46-4 | 99.1% | 500 MG
SDP

Catalog No. 5000409422
Encompass_Preferred

Suprofen is a non-steroidal anti-inflammatory drug (NSAID). It acts as an ibuprofen-type analgesic and antipyretic by inhibiting prostaglandin synthesis, and has been proposed for anti-arthritic applications.

  • Inhibits prostaglandin synthesis
  • Non-steroidal anti-inflammatory drug
  • Ibuprofen-type analgesic and antipyretic
  • Target is PGE synthase
  • Appears as a white to off-white solid
  • Molecular weight of 260.31
  • Chemical formula is C14H12O3S

Catalog No. 50-004-09422 Supplier Medchemexpress LLC Supplier No. HYB0270500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suprofen is a non-steroidal anti-inflammatory drug (NSAID). It acts as an ibuprofen-type analgesic and antipyretic by inhibiting prostaglandin synthesis, and has been proposed for anti-arthritic applications.

  • Inhibits prostaglandin synthesis
  • Non-steroidal anti-inflammatory drug
  • Ibuprofen-type analgesic and antipyretic
  • Target is PGE synthase
  • Appears as a white to off-white solid
  • Molecular weight of 260.31
  • Chemical formula is C14H12O3S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.