Learn More
Electron Microscopy Sciences Superslip Micro Glasses #1, 24x60 mm, 54 pieces/oz package 1 OZ
SDP
Catalog No. 501899799
Encompass_Preferred
Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These "Superslip" coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run. These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.
Size: 24x60 mm
Package: 54 pieces/oz
Catalog No. 50-189-9799 Supplier Electron Microscopy Sciences Supplier No. 7223660
Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These "Superslip" coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run. These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.
Size: 24x60 mm
Package: 54 pieces/oz