Learn More
Electron Microscopy Sciences Superslip Micro Glasses #1, 24x40 mm, 80 pieces/oz package 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7223641
Encompass_Preferred
Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These "Superslip" coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run. These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.
Size: 24x40 mm
Package: 80 pieces/oz
Catalog No. 50-189-9796