Learn More
Aviva Systems Biology Corp SUPT16H Antibody - middle region (ARP38921_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38921_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUPT16H. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUPT16H. Alternate names: CDC68, FACT, FACTP140, FLJ10857, FLJ14010, FLJ34357, SPT16/CDC68ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56849