Learn More
Aviva Systems Biology Corp SUPT3H Antibody - N-terminal region (ARP30038_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30038_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SUPT3H. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SUPT3H. Alternate names: SPT3, SPT3LApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54107