Learn More
Aviva Systems Biology Corp SUPT5H Antibody - middle region (ARP34064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUPT5H. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUPT5H. Alternate names: FLJ34157, SPT5, SPT5H, Tat-CT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55235