Learn More
Aviva Systems Biology Corp SUPT5H Antibody - C-terminal region (ARP33386_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33386_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUPT5H. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SUPT5H. Alternate names: SPT5, SPT5H, Tat-CT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Zebrafish
Catalog No. 50-014-54992