Learn More
Aviva Systems Biology Corp SUPT6H Antibody - N-terminal region (ARP38260_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38260_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SUPT6H. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SUPT6H. Alternate names: SPT6, SPT6H, emb-5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53102