Learn More
Aviva Systems Biology Corp SUPV3L1 Antibody - N-terminal region (ARP36353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUPV3L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SUPV3L1. Alternate names: SUV3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56124