Learn More
AdipoGen Suramin Hexasodium salt, 129-46-4, MFCD00210217, 50 mg
SDP
Catalog No. 501146548
Encompass_Preferred
MF: C51H34N6O23S6 . 6Na, MW: 1429.180, ≥98%, Appearance: White to off-white powder. Potent ATPase inhibitor. Potent competitive inhibitor of reverse transcriptase. Shows anti-HIV activity. Anticancer compound. Protein kinase C (PKC) inhibitor.
Catalog No. 50-114-6548 Supplier AdipoGen Supplier No. AGCR13575M050
MF: C51H34N6O23S6 . 6Na, MW: 1429.180, ≥98%, Appearance: White to off-white powder. Potent ATPase inhibitor. Potent competitive inhibitor of reverse transcriptase. Shows anti-HIV activity. Anticancer compound. Protein kinase C (PKC) inhibitor.