Learn More
Medchemexpress LLC Suramin sodium salt | 129-46-4 | 100.0% | 10 MG
SDP
Catalog No. 5000296701
Encompass_Preferred
Suramin sodium salt is the hexasodium salt of suramin provided for research use as a reversible and competitive protein-tyrosine phosphatase inhibitor. It is offered as a high-purity solid with documented solubility in DMSO and water and defined storage recommendations for both solid and solution states.
  • High purity solid (99.98%).
  • Soluble in DMSO and water (50 mg/mL; may require ultrasonic or warming).
  • Supplied as a 10 mg package suitable for small-scale studies.
  • Intended for enzyme inhibition and pharmacology research applications.
  • Store solid at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-96701 Supplier Medchemexpress LLC Supplier No. HYB0879A10MG
Suramin sodium salt is the hexasodium salt of suramin provided for research use as a reversible and competitive protein-tyrosine phosphatase inhibitor. It is offered as a high-purity solid with documented solubility in DMSO and water and defined storage recommendations for both solid and solution states.
  • High purity solid (99.98%).
  • Soluble in DMSO and water (50 mg/mL; may require ultrasonic or warming).
  • Supplied as a 10 mg package suitable for small-scale studies.
  • Intended for enzyme inhibition and pharmacology research applications.
  • Store solid at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month).