Learn More
Medchemexpress LLC Suramin sodium salt | 129-46-4 | 100.0% | 25 MG
SDP
Catalog No. 5000450652
Encompass_Preferred
Suramin sodium salt is a reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor. It also acts as a potent inhibitor of sirtuins (SirT1, SirT2, and SirT5) and a competitive inhibitor of reverse transcriptase (DNA topoisomerase II). Additionally, this agent is a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor, efficiently inhibits IP5K, and functions as an antiparasitic, anti-neoplastic, and anti-angiogenic agent, intended for research use only.
  • Reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor.
  • Potent inhibitor of sirtuins (SirT1, SirT2, and SirT5).
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II).
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor.
  • Efficiently inhibits IP5K.
  • Acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent.
  • Intended for research use only.
Catalog No. 50-004-50652 Supplier Medchemexpress LLC Supplier No. 25MGHYB0879A
Suramin sodium salt is a reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor. It also acts as a potent inhibitor of sirtuins (SirT1, SirT2, and SirT5) and a competitive inhibitor of reverse transcriptase (DNA topoisomerase II). Additionally, this agent is a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor, efficiently inhibits IP5K, and functions as an antiparasitic, anti-neoplastic, and anti-angiogenic agent, intended for research use only.
  • Reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor.
  • Potent inhibitor of sirtuins (SirT1, SirT2, and SirT5).
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II).
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor.
  • Efficiently inhibits IP5K.
  • Acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent.
  • Intended for research use only.