Learn More
Medchemexpress LLC Suramin sodium salt | 129-46-4 | 100.0% | 5 MG
SDP
Catalog No. 5000409162
Encompass_Preferred
Suramin sodium salt is a versatile compound acting as a reversible and competitive inhibitor of protein-tyrosine phosphatases (PTPases). It also inhibits sirtuins and reverse transcriptase. It has been identified as an antiparasitic, anti-neoplastic, and anti-angiogenic agent, and is also a potent inhibitor of SARS-CoV-2 RNA-dependent RNA polymerase (RdRp).
  • Reversible and competitive PTPases inhibitor
  • Potent inhibitor of sirtuins (SirT1, SirT2, SirT5)
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II)
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor
  • Efficiently inhibits IP5K
  • Acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent
Catalog No. 50-004-09162 Supplier Medchemexpress LLC Supplier No. HYB0879A5MG
Suramin sodium salt is a versatile compound acting as a reversible and competitive inhibitor of protein-tyrosine phosphatases (PTPases). It also inhibits sirtuins and reverse transcriptase. It has been identified as an antiparasitic, anti-neoplastic, and anti-angiogenic agent, and is also a potent inhibitor of SARS-CoV-2 RNA-dependent RNA polymerase (RdRp).
  • Reversible and competitive PTPases inhibitor
  • Potent inhibitor of sirtuins (SirT1, SirT2, SirT5)
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II)
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor
  • Efficiently inhibits IP5K
  • Acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent