Learn More
Medchemexpress LLC Suramin sodium salt | 129-46-4 | 99.98% | 100 MG
SDP
Catalog No. 5000408606
Encompass_Preferred
Suramin sodium salt is a reversible and competitive inhibitor of protein-tyrosine phosphatases (PTPases). It also inhibits sirtuins (SirT1, SirT2, and SirT5), acts as a competitive inhibitor of reverse transcriptase (DNA topoisomerase II), and is a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor. Additionally, it efficiently inhibits IP5K, functioning as an antiparasitic, anti-neoplastic, and anti-angiogenic agent.
  • Reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor
  • Inhibits sirtuins (SirT1, SirT2, SirT5)
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II)
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor
  • Efficiently inhibits IP5K
  • Acts as an antiparasitic agent
  • Acts as an anti-neoplastic agent
  • Acts as an anti-angiogenic agent
Catalog No. 50-004-08606 Supplier Medchemexpress LLC Supplier No. HYB0879A100MG
Suramin sodium salt is a reversible and competitive inhibitor of protein-tyrosine phosphatases (PTPases). It also inhibits sirtuins (SirT1, SirT2, and SirT5), acts as a competitive inhibitor of reverse transcriptase (DNA topoisomerase II), and is a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor. Additionally, it efficiently inhibits IP5K, functioning as an antiparasitic, anti-neoplastic, and anti-angiogenic agent.
  • Reversible and competitive protein-tyrosine phosphatases (PTPases) inhibitor
  • Inhibits sirtuins (SirT1, SirT2, SirT5)
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II)
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor
  • Efficiently inhibits IP5K
  • Acts as an antiparasitic agent
  • Acts as an anti-neoplastic agent
  • Acts as an anti-angiogenic agent