Learn More
Medchemexpress LLC Suramin (sodium salt) | 129-46-4 | MFCD00003780 | 100.0% | 50 MG
SDP
Catalog No. 5000409339
Encompass_Preferred
Suramin sodium salt is a reversible and competitive inhibitor of protein-tyrosine phosphatases (PTPases). It also potently inhibits sirtuins (SirT1, SirT2, SirT5), acts as a competitive inhibitor of reverse transcriptase (DNA topoisomerase II), and functions as a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor. Additionally, it efficiently inhibits IP5K and acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent.
  • Reversible and competitive PTPases inhibitor
  • Potent inhibitor of sirtuins (SirT1, SirT2, SirT5)
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II)
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor
  • Efficiently inhibits IP5K
  • Acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent
Catalog No. 50-004-09339 Supplier Medchemexpress LLC Supplier No. HYB0879A50MG
Suramin sodium salt is a reversible and competitive inhibitor of protein-tyrosine phosphatases (PTPases). It also potently inhibits sirtuins (SirT1, SirT2, SirT5), acts as a competitive inhibitor of reverse transcriptase (DNA topoisomerase II), and functions as a potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor. Additionally, it efficiently inhibits IP5K and acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent.
  • Reversible and competitive PTPases inhibitor
  • Potent inhibitor of sirtuins (SirT1, SirT2, SirT5)
  • Competitive inhibitor of reverse transcriptase (DNA topoisomerase II)
  • Potent SARS-CoV-2 RNA-dependent RNA polymerase (RdRp) inhibitor
  • Efficiently inhibits IP5K
  • Acts as an antiparasitic, anti-neoplastic, and anti-angiogenic agent