Learn More
AdipoGen Suramin sodium salt
SDP
Catalog No. 502562279
Encompass_Preferred
Chemical. CAS 129-46-4. Formula C51H34N6O23S6 . 6Na. MW 1291.2 . 137.9. Potent ATPase inhibitor. Potent competitive inhibitor of reverse transcriptase. Shows anti-HIV activity. Anticancer compound. Protein kinase C PKC inhibitor. Potent inhibitor of melanoma heparanase and tumor cell metastasis. Non-specific growth factors inhibitor including PDGF, EGF, aFGF and bFGF. TGF-beta1 inhibitor. Topoisomerase I and II inhibitor. Interleukin-1 IL-1 inhibitor. Interleukin-4 IL-4 inhibitor. G protein inhibitor. P2X and P2Y purinergic receptor antagonist. Antiangiogenic. Potent VEGF inhibitor. Telomerase inhibitor. Shows adjuvant properties. Regulates ryanodine receptor. Direct adenylyl cyclase inhibitor. Protein synthesis inhibitor. SIRT1 sirtuin 1 and SIRT5 sirtuin 5 inhibitor. Immunosuppressive. Antifibrotic agent. Antiparasitic. Antiprotozoal. Athelmintic.
Catalog No. 50-256-2279 Supplier AdipoGen Supplier No. AGCR13575VM250
Chemical. CAS 129-46-4. Formula C51H34N6O23S6 . 6Na. MW 1291.2 . 137.9. Potent ATPase inhibitor. Potent competitive inhibitor of reverse transcriptase. Shows anti-HIV activity. Anticancer compound. Protein kinase C PKC inhibitor. Potent inhibitor of melanoma heparanase and tumor cell metastasis. Non-specific growth factors inhibitor including PDGF, EGF, aFGF and bFGF. TGF-beta1 inhibitor. Topoisomerase I and II inhibitor. Interleukin-1 IL-1 inhibitor. Interleukin-4 IL-4 inhibitor. G protein inhibitor. P2X and P2Y purinergic receptor antagonist. Antiangiogenic. Potent VEGF inhibitor. Telomerase inhibitor. Shows adjuvant properties. Regulates ryanodine receptor. Direct adenylyl cyclase inhibitor. Protein synthesis inhibitor. SIRT1 sirtuin 1 and SIRT5 sirtuin 5 inhibitor. Immunosuppressive. Antifibrotic agent. Antiparasitic. Antiprotozoal. Athelmintic.