Learn More
Selleck Chemical LLC Suramin sodium salt-5mg
SDP
Catalog No. 502390035
Encompass_Preferred
Suramin sodium salt (Suramin hexasodium salt) is a reversible and competitive protein-tyrosine phosphatase (PTPases) inhibitor, also is the potent inhibitor of sirtuins, purified topoisomerase II and SARS-CoV-2 RNA-dependent RNA polymerase (RdRp).
Catalog No. 50-239-0035 Supplier Selleck Chemical LLC Supplier No. S89425MG
Suramin sodium salt (Suramin hexasodium salt) is a reversible and competitive protein-tyrosine phosphatase (PTPases) inhibitor, also is the potent inhibitor of sirtuins, purified topoisomerase II and SARS-CoV-2 RNA-dependent RNA polymerase (RdRp).