Learn More
Sigma Aldrich Fine Chemicals Biosciences Suramin sodium salt 98 TLC25MG
Catalog No. 502899916
Encompass
Suramin is an antiparasitic drug which blocks endothelial cell proliferation and increases tyrosine phosphorylation of many signalling proteins. It is associated with urticaria paresthesias vomitingnbspand neurologic toxicity. Suramin is used to treat trypanosomiasis and onchocerciasis. It functions as an inhibitor of heparanasenbspand also exhibits anti-AIDS (acquired immunodeficiency syndrome) property.
Catalog No. 50-289-9916 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S267125MG
Suramin is an antiparasitic drug which blocks endothelial cell proliferation and increases tyrosine phosphorylation of many signalling proteins. It is associated with urticaria paresthesias vomitingnbspand neurologic toxicity. Suramin is used to treat trypanosomiasis and onchocerciasis. It functions as an inhibitor of heparanasenbspand also exhibits anti-AIDS (acquired immunodeficiency syndrome) property.